General-Purpose TEFC/ODP, NEMA Type C-Face Four-In-One Motors
Fournisseur: PALM ELECTRIC MOTORS & SUPPLIES
Certificats
A propos de cet article
Totally enclosed motors are ideal for dusty, dirty, or damp environments.
- Unique four-in-one design allows for replacement of ODP, TEFC, C-Face, or rigid based motors
- Use for vertical or horizontal applications
Single-phase motors include a removable, rigid base. Three-phase motors inlcude a removable base.
Spécifications
- Catalog No:
- MFLX71004-25
- MFLX71005-15
- Motor enclosure type:
- TEFC, ODP
- TEFC, ODP
- Power conduit box connection:
- Yes
- Yes
- Warranty:
- 1 year
- 1 year