Cast iron float and thermostatic steam traps (parallel configuration)
Fournisseur: SPIRAX SARCO INC
Certificats
A propos de cet article
Keep steam space free of condensate to provide maximum system efficiency.
- Constructed of durable cast iron
Float and thermostatic steam traps contain a float valve mechanism which modulates to discharge condensate continuously at steam temperature, while non-condensable gases are released by a separate internal balanced pressure thermosatic air vent. Stainless steel internal parts can be serviced without disturbing the piping connections.
Typical applications:
All process equipment, particularly when controlled by modulating temperature control valves
Unit heaters
Air heat coils
Heat exchangers
Steam main drip stations
Spécifications
- Catalog No:
- MFLX07600-30
- Max. operating pressure:
- 15 psi
- Max. temperature:
- 232 °C of superheat (450 °F of superheat)
- Warranty:
- 1 year