Plastic Hand-Operated Water and Chemical Siphon/Drum Pump
Fournisseur: BECKSON INDUSTRIAL PRODUCTS
Certificats
A propos de cet article
Ideal for most acid/base or ionic salt applications.
- Portable and self-priming
These plastic hand-operated drum pumps are ideal for water or chemical siphons. All pumps are constructed with PVC barrel and hose. Water siphon model 70607-00 includes a 60” PVC outlet hose that can be cut to fit your application. Chemical siphon models 70607-10 and 70607-12 are suitable for acid and alkali transfer.
Spécifications
- Catalog No:
- MFLX70607-10
- MFLX70607-12
- MFLX70607-00
- Flow rate:
- 7 strokes/L (26 strokes/gal.)
- 4 strokes/L (12 strokes/gal.)
- 9 strokes/L (32 strokes/gal.)
- Tube ext.Ø, outlet:
- 1.4 cm (0.5625")
- 1.4 cm (0.5625")
- 1.4 cm (0.5625")
- Tube length, inlet:
- 45.7 cm (18")
- Tube length, outlet:
- 121.9 cm (48")
- 121.9 cm (48")
- 152.4 cm (60")
- Wetted materials:
- PVC barrel and hose, LDPE tube
- PVC barrel and hose, LDPE tube
- PVC barrel, shaft, hose
- Warranty:
- None
- None
- None