Chemical-Duty Self-Priming Transfer Pumps
Fournisseur: MARCH MANUFACTURING INC
Certificats
A propos de cet article
Different wetted materials for different chemical compatibilities.
- Pumps with glass-filled polypropylene heads are ideal for transferring a wide range of caustic materials
- Pumps with virgin PVDF heads are ideal for DI H2O, strong acids, and halogens
Self-priming pumps with glass-filled polypropylene heads are capable of lifting water up to 10 feet.
Delivery: Includes conduit box for hardwiring.
Spécifications
- Catalog No:
- MFLX07190-60
- MFLX07190-20
- Duty cycle continuous:
- Yes
- Yes
- Inlet connection:
- 1-1/2" NPT(F)
- 1" NPT(F)
- Max. pressure:
- 50 psi
- 75 psi
- Motor hp:
- 1
- 1/2
- Outlet connection:
- 1" NPT(M)
- 3/4" NPT(M)
- Power:
- 115/230 VAC; 50/60 Hz; 11.8/5.9 A; 1 phase
- 115/230 VAC; 50/60 Hz; 7.4/3.7 A; 1 phase
- Wetted materials:
- Glass-filled polypropylene, 99,9% aluma oxide ceramic, carbon brushing, Viton®
- Carbon-filled Kynar®, 99,9% aluma oxide ceramic, carbon brushing, Viton®