LMI Solenoid-Diaphragm Metering Remote Control Pumps
Fournisseur: EQUIP SOLUTIONS
Certificats
A propos de cet article
Manually adjustable stroke length and stroke frequency provide precise control of flow rates.
- Clear face cover protects pump controls on low flow models (1 GPH and lower)
- Chemical-resistant housing provides protection in harsh environments
An external pulse trigger can be accepted for remote stroke frequency control. Low-flow pump models (1 GPH and lower) include easy access to 4 pin connections.
Service kits include a replacement diaphragm, seal, four pump head screws, and check valves.
Delivery: Includes four-function valve, 16-ft polyethylene tubing, foot valve/strainer, and 8-ft power cord with molded plug.
Spécifications
- Catalog No:
- MFLX74514-40
- MFLX74514-50
- MFLX74514-20
- Accuracy:
- ±2%
- ±2%
- ±2%
- CSA regulatory approval:
- Yes
- Yes
- Yes
- Inlet connection:
- 3/8" OD tubing
- 1/2" OD tubing
- 3/8" OD tubing
- Max. pressure:
- 60 bar (60 psi)
- 25 psi
- 100 bar (100 psi)
- Min. flow rate:
- 2.52 ml/min (0.45 gal./h)
- 6.33 ml/min (0.1 gal./h)
- 0.79 ml/min (0.013 gal./h)
- Outlet connection:
- 3/8" OD tubing
- 1/2" OD tubing
- 3/8" OD tubing
- Wetted materials:
- Ceramic, Fluorofilm®, PVC, PVDF/Polyprel®
- Ceramic, Fluorofilm® , PTFE, PVC
- Ceramic, Fluorofilm®, PVC, PVDF/Polyprel®