Burkert® Micro Solenoid Rocker Valves
Fournisseur: BURKERT FLUID CONTROL SYSTEMS
Certificats
A propos de cet article
No worrying about reliability—longest life available.
- Eliminates heat transfer from diaphragm to actuator
- Easy purging—minimal internal volume
- Ideal for manifold mounting
Rocker Valves have a miniature rocker that operates the isolated diaphragm to separate actuator from the media. Valve is only 16 mm wide. Each valve is equipped with an LED display and push-on tube connections. Valves include two 19" electrical leads.
Spécifications
- Catalog No:
- MFLX98622-00
- Actuation time:
- 30 ms
- Max. pressure:
- 28 psi
- Max. temperature:
- 55 °C (131 °F)
- Pilot valve air connection:
- 0.25-28 UNF
- Power:
- 24 VDC. 3.4 W
- Wetted materials:
- PVDF body, FFKM seals
- Warranty:
- 18 months