Double-Walled Heat-Shrink Tubing, PTFE
Fournisseur: VWR International
Certificats
A propos de cet article
Ideal for chemically inert applications.
- Global market leader in range of formulations for most extensive chemical compatibility
Double-walled PTFE tubing combines an exterior of heat-shrink PTFE with an inner layer of FEP to provide a tight, moisture-proof bond. Comes in 4-ft lengths; 3 lengths per pack, for a total of 12 ft/pk. Opaque.
Spécifications
- Catalog No:
- MFLX95810-15
- MFLX95810-13
- MFLX95810-16
- Clarity:
- Opaque
- Opaque
- Opaque
- Length per pack:
- 3.7 m (12 ft.)
- 3.7 m (12 ft.)
- 3.7 m (12 ft.)
- Warranty:
- Out of box
- Out of box
- Out of box