Hand-operated siphon/lift drum pump, 16 strokes/gallon
# de catalogue MFLX06437-10
Fournisseur: BECKSON INDUSTRIAL PRODUCTS
Spécifications
- Description:Hand-operated siphon/lift drum pump, 16 strokes/gallon
- Wetted materials:PVC body and hose; Hypalon® valves and seals; LDPE tube
- Inlet connection:2" IPS
- Tube ext.Ø, outlet:1.25"
- Tube length, inlet:18"
- Tube length, outlet:18"
- Warranty:1 month
- Cat. no.:MFLX06437-10
Spécifications
A propos de cet article
Ideal for most acid/base or ionic salt applications.
- Equipped with a 2" IPS bung adapter for pumping detergents, water, dilute alkalies, or waxes from drums
- Cut inlet tube to fit your container
This plastic hand-operated drum pump is ideal for pumping detergents, water, dilute alkalies, or waxes from drums. Constructed with a PVC body and hose, Hypalon valves and seals, and an LDPE tube. This pump offers 16 strokes/gallon (5 strokes/liter).
- Description:Hand-operated siphon/lift drum pump, 16 strokes/gallon
- Wetted materials:PVC body and hose; Hypalon® valves and seals; LDPE tube
- Inlet connection:2" IPS
- Tube ext.Ø, outlet:1.25"
- Tube length, inlet:18"
- Tube length, outlet:18"
- Warranty:1 month
- Cat. no.:MFLX06437-10
Customers Who Bought This Also Bought