Hand-Operated Piston Dispensing Drum Pump, Polypropylene
Supplier: Action Pump
Certificates
About this item
Economical piston pumps are ideal for a variety of fluids.
- Adaptable to most pails and drums
Models 06512-00 and -05 have adjustable strokes to allow for dispensing different amounts fo fluid with just one pump. Model 06512-00 pumps 64, 32, 22 and 16 strokes per gallon, model 60512-05 pumps 32, 16, 11 and 8 strokes per gallon.
Specifications
- Catalog No:
- MFLX06512-00
- MFLX06512-05
- Max. temperature:
- 82 °C (180 °F)
- 82 °C (180 °F)
- Max. viscosity:
- 4000 SSU
- 4000 SSU
- Outlet connection:
- Spout
- Spout
- Tube ext.Ø, inlet:
- 1.27 cm (0.5")
- 1.27 cm (0.5")
- Tube ext.Ø, outlet:
- 1.27 cm (0.5")
- 1.27 cm (0.5")
- Tube length, inlet:
- 81 cm (32")
- 81 cm (32")
- Wetted materials:
- Polypropylene, Polyethylene
- Polypropylene, Polyethylene
- Warranty:
- 1 year
- 6 months