American Machine & Tool Motor Foot Bracket for NEMA Type 56 C-Face Motors
# de catalogue MFLX07127-80
Fournisseur: AMT PUMP COMPANY
Spécifications
- Description:Motor foot bracket, provides base for NEMA type 56 C-face motors
- Warranty:1 year
- Cat. no.:MFLX07127-80
- Motor Frame Size:56C
- Length:15.24 cm
- Width:7.62 cm
Spécifications
A propos de cet article
Create your own motor mounting with the hardware below.
- Easily mount your American Machine & Tool Motor NEMA Type 56 C-face motors
Motor Foot Bracket provides a base for NEMA Type 56 C-face motors.
- Description:Motor foot bracket, provides base for NEMA type 56 C-face motors
- Warranty:1 year
- Cat. no.:MFLX07127-80
- Motor Frame Size:56C
- Length:15.24 cm
- Width:7.62 cm
Customers Who Bought This Also Bought