Coupling hardware for 5/8" diameter bore, 7.2 hp maximum
# de catalogue MFLX07127-59
Fournisseur: VWR International
Spécifications
- Description:Coupling hardware for 5/8" diameter bore, 7,2 hp maximum
- Warranty:None
- Cat. no.:MFLX07127-59
- Motor HP:7.2
Spécifications
A propos de cet article
Create your own motor mounting with the hardware below.
- Optimize performance of your Pump Motor
Order one coupling with the same diameter as the motor driving shaft, one coupling with the same diameter as the driven shaft, and one spider (see accessories).
- Description:Coupling hardware for 5/8" diameter bore, 7,2 hp maximum
- Warranty:None
- Cat. no.:MFLX07127-59
- Motor HP:7.2
Customers Who Bought This Also Bought