GF Signet PVC Saddle Installation Fittings
Fournisseur: BURT PROCESS EQUIPMENT
Certificats
A propos de cet article
Ensure proper insertion depth and measurement.
- Constructed of durable PVC 40 or PVC 80
These PVC saddle installation fittings are available for PVC 40 or PVC 80 pipes from 2” to 8” diameters.
Spécifications
- Connection type:
- Saddle
- Saddle
- Saddle
- Saddle
- Orifice dimensions:
- 2"
- 2.5"
- 3"
- 6"
- Warranty:
- 1 year part and 2 years labor
- 1 year part and 2 years labor
- 1 years part and 2 years labor
- 1 year part and 2 years labor