Burkert Paddle-Wheel Sensors with Integral Fittings
Fournisseur: BURKERT FLUID CONTROL SYSTEMS
Certificats
A propos de cet article
Enhance accuracy of your flow readings.
- Can be used with various liquids
These paddle-wheel sensors are ideal for measuring both neutral and aggressive, solid-free liquids. Built-in pipe fittings ensure simple installation into pipes from 1/2" up to 2". Mount sensor/fittings horizontally or vertically.
Choose from polyvinyl chloride (PVC), brass, or stainless steel (SS) sensor/fittings. PVC models come with true union fittings (solvent/fusion connection). Order brass and SS models with NPT(F) fittings for high temperature or high pressure applications. SS models are especially ideal for measuring aggressive fluids. Wetted parts are polyvinylidene fluoride, ceramic, fluoropolymer, and PVC, brass, or SS.
Spécifications
- Catalog No:
- MFLX33117-15
- Max. pressure:
- 235 psi
- Max. operating temperature:
- 50 °C (122 °F)
- Repeatability:
- ±0.4% of reading
- Accuracy:
- ±1% of reading
- Warranty:
- 1 year