VWR® High Capacity Magnetic Stirrers
Supplier: VWR International
Certificates
About this item
VWR® high capacity magnetic stirrers with digital speed control and ceramic coated top plate.
- Powerful motor drive for high volume stirring
- Coated white ceramic top plate
- Digital speed control
VWR® high capacity magnetic stirrers with digital speed control and ceramic coated top plate. Powerful motor drive capable of stirring volumes up to 50/150 L.
Certifications: CE marked.
Specifications
- Catalog No:
- 76549-944
- Timer:
- 1 min to 9 h
- W×D×H: