VWR® Mini Dry Bath Block Heater
76549-942
- Temperature Range:RT 5...100 °C
- Capacity:15× 1.5/2.0 or 2× 50 ml
- Description:VWR® mini dry bath block heater
- Cat. no.:76549-942
- W×D×H:11.7×15×11.2 cm (4.6×5.9×4.4")
- Electrical:100/240 V, 50/60 Hz, 50 W
- Temperature Accuracy:±0.5 °C
VWR® mini dry bath block heater with digital temperature control up to 100 °C.
- Digital temperature control
- Wide range of block accessories for all common tubes and plates
- Extremely compact, only 4.6" wide
- Temperature increment: 0.1 °C
The unit includes a hinged lid, improving temperature uniformity throughout tube samples. With a wide range of accessory blocks, the instrument is ideal for incubating tubes from 0.2 to 50 ml.
Certifications: CE marked.
Ordering information: Heaters require a VWR® modular heating block for operation (sold separately).
Customers Who Bought This Also Bought