Ward's® Corn Meal Agar
Fournisseur: VWR International
Certificats
A propos de cet article
Medium for cultivating phytopathological fungi, and stimulating production of chlamydospores.
- Sterile, no autoclaving
- Conveniently prepared for you in both Agar Plates and Agar Slants
- General-purpose medium used to cultivate yeasts and molds
Corn Meal Agar is valuable for morphologic differentiation of many yeast-like organisms. It suppresses vegetative growth of many fungi while stimulating sporulation. Corn Meal Agar allows Candida albicans to produce chlamydospores which is one of the best criterion for identification.
Delivery: Prepared media will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Caution: For educational use only.
Customers Who Bought This Also Bought