VWR® Magnetic Hotplate Stirrers
Supplier: VWR International
Certificates
About this item
VWR® magnetic hotplate stirrers, with temperature control up to 400 °C and stirring speeds up to 1500 rpm.
- White ceramic surface for excellent chemical resistance
- Digital speed control up to 400 °C
- Available with external temperature probe (displays actual sample temperature)
- 'HOT' LED safety indicator alerts user when plate is unsafe to touch
Corresponding magnetic stirrers (stirring only 76549-916 and 76549-924) or hotplates (heating only 76549-914 and76549-922) with ceramic plates are also available. Full specification details are given under separate entries, please view full details by searching for the relevant product code.
Certifications: CE marked.
Specifications
- Catalog No:
- 76549-918
- 76549-912
- Control type:
- Digital
- Digital