Cole-Parmer® SH-200 Series Microplate Shakers, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Provides a fast-mixing action in a tight orbit.
- Non-slip mat and platform lip securely hold plates
- Built-in timer and digital speed LED display provide reproducible results
- Can be use in incubators
These microplate shakers combine high speed and a tiny 1.5 mm orbit to produce vigorous shaking perfect for microwell plates. The shakers offer a quiet drive mechanism, easy to use controller with timer, LED display, and a digital speed selection from 250 to 1250 rpm with a soft start.
Easily set digital timer from 1 to 999 minutes. When timer reaches zero, the shaker automatically stops and sounds an alert. Shaker is capable of continuous use, and the microprocessor maintains speed accuracy over long runs.
Choose the four-plate model for small scale applications or the eight-plate model for high throughput bench_top applications.
Delivery: Supplied with flat platform, non-slip mat and power cord.
Spécifications
- Catalog No:
- 76542-136
- 76542-134
- Model:
- SH-200D-M-120
- SH-200D-M-L-120
- Max. load:
- 1 kg (2.2 lbs.)
- 2 kg (4.4 lbs.)
- Operating temperature range:
- 4...40 °C (39.2...104 °F)
- 4...40 °C (39.2...104 °F)
- Display:
- LED
- LED