Cole-Parmer®Large Capacity Digital Stirring Hot Plates, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Easy to use temperature and stirring control dials.
- Large square plate area ideal for heating one large vessel or several smaller ones
- Hot warning light, remains active even when unplugged while above 50 °C
- Independent safety circuit to protect against overheating
Temperature and stirring parameters are controlled by easy-to-use dials. Each model features a ‘Hot’ warning light which flashes whenever the plate temperature is above 50 °C. Powerful stirring action allows for stirring volumes up to 15 Liters. Rugged metal heats quickly and has even temperature distribution across the entire plate. Chemical-resistant ceramic stays feature higher max temperatures.
Customers Who Bought This Also Bought