Programmable Refrigerated Eco Incubator, INE Series, Yamato
Fournisseur: Yamato Scientific
Certificats
A propos de cet article
Upgraded inverter control improved refrigeration efficiency, reduced frost significantly and minimized wasted power during refrigeration.
- 44% power savings compared to previous models
- Controller upgraded for easier viewing and operability
- Temperature distribution accuracy improved for better incubation
Standard equipped with program operation, auto-stop, auto-start, quick auto stop, timer, calibration off-set, memory, electricity & CO₂ emission monitor.
Spécifications
- Catalog No:
- 76550-248
- 76550-246
- Temperature setting accuracy:
- ±0.5 °C (at 37 °C during continuous operation)
- ±0.5 °C (at 37 °C during continuous operation)
- Temperature range:
- –5…65 °C
- –5…65 °C
- Shelf weight capacity:
- 15 kg/pc
- 15 kg/pc
- Electrical:
- 220 VAC, 50/60 Hz, 4.5 A
- 115 VAC, 50/60 Hz, 8.7 A