AssureWear™ High Top Boot Covers, Medicom
# de catalogue CA76545-406
Fournisseur: Medicom
Spécifications
- Clothing Size:XL
- Garment application:Industrial
- Gender:Unisex
- Color:Blue
- Cat. no.:CA76545-406
Spécifications
A propos de cet article
Footwear protection with fluid-repellent, slip-on covers.
- Non-woven fabric sewn in the shape of a boot
- Elastic on toes, heel, and ankle
- Single use
- Latex-free
High top boot covers intended to be worn by operating room personnel during various procedures to protect patient/materials and the operating personnel from transfer of microorganisms, particulates, etc.
Boot covers are made from non-woven fabric sewn in the shape of a boot, with elastic on the top of the toes, behind the heel, and at the ankle. They feature reinforced fabric from the sole up to the ankle (glossy side inside the boot cover) and 3 lines on the sole.
Packaging: Boot covers are packaged 50/box, 3 boxes/case.
Customers Who Bought This Also Bought