SafeMask® Architect Pro™ Surgical/N95 Respirator, Medicom
Fournisseur: Medicom
Certificats
A propos de cet article
Medical grade respirator providing optimal respiratory protection by filtering 95% of particulate aerosols free of oil.
- High-quality materials and duck-bill shape
- Lightweight, breathable construction
- Unique single split comfort band
- Adjustable nose piece
This medical grade respirator has been evaluated for use in healthcare settings according to recognized standards for biocompatibility, flammability, and fluid resistance. It provides optimal respiratory protection by filtering 95% of particulate aerosols free of oil. High-quality materials and duckbill shape offer optimal breathability, with no exhalation valve needed. Unique single split comfort bands hold mask in place without pulling or irritating skin. Lightweight construction ensures comfort and adjustable nose piece creates a personalized fit. Convenient 8-hour wear time.
Packaging: Masks are packaged 50/box, 10 boxes/case.
Customers Who Bought This Also Bought