Spécifications
- Description:Eutectic alloys
- Cat. no.:470338-588
- Includes:Instructional manual and SDS booklet
- No. of Uses:5
Spécifications
A propos de cet article
Create your own metal eutectic alloy - a combination of two metals that has a lower melting point that the melting point of either metal.
- Understand and demonstrate the formation of metal eutectic alloys
- Aligned to NGSS Practices
Form a metal alloy as bismuth and tin are heated to the eutectic point of the two metals which is lower than the melting point of either metal.