Purifier® Vertical Clean BenchVertical, 6', Labconco®
Fournisseur: Labconco
Certificats
A propos de cet article
Purifier Vertical Clean Benches direct HEPA-filtered air downward over the work area to provide ISO Class 5 conditions and protect your work from particulate contamination.
Used in pharmacy compounding, plant tissue culture, media plate preparation, electronics inspection, medical device assembly and more.
Certifications: Promotes ISO Class 5 air (formerly Class 100 per Federal Standard 209E)
BodyTextAccessories: The IV Bar supports intravenous solution bottles and bags. Includes IV bar, mounting hardware, four hangers and instructions for installation.
Spécifications
- Catalog No:
- 76545-620
- 76545-614
- 76545-612
- 76545-618
- 76545-616
- 76545-610
- 76545-624
- 76545-622
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Ext. W×D×H:
- 182.88×73.66×116.84 cm (72×29×46")
- 182.88×73.66×116.84 cm (72×29×46")
- 182.88×73.66×116.84 cm (72×29×46")
- 182.88×73.66×116.84 cm (72×29×46")
- 182.88×73.66×116.84 cm (72×29×46")
- 182.88×73.66×116.84 cm (72×29×46")
- 182.88×73.66×116.84 cm (72×29×46")
- 182.88×73.66×116.84 cm (72×29×46")
- Environmental certification:
- 70% Recycled Content
- 70% Recycled Content
- 70% Recycled Content
- 70% Recycled Content
- 70% Recycled Content
- 70% Recycled Content
- 70% Recycled Content