Sentinel XL® CBRN System, with Rechargeable NIMH Battery and Duffel, ILC Dover
# de catalogue 76533-628
Fournisseur: ILC DOVER LP
Spécifications
- Description:Sentinel XL® CBRN system
- Size:Universal
Spécifications
A propos de cet article
The Sentinel XL® CBRN PAPR system, which features an adjustable headband, is comfortable to wear and requires no fit-testing. The “lay flat” design protects the system and allows it to stow easily in a duffel bag.
- Lightweight duffel bag included for easy transport
- NiMH recharable battery included
- Adjustable anti-microbial headband
- NIOSH approved at CAP 2 rating
- Latex-free
The Sentinel XL® CBRN PAPR system includes Butyl Hood, Sentinel XL® Blower, NiMH battery with charging cord, 3 CBRN cartridge filters, and a duffel bag.
Certifications: NIOSH approved.
Customers Who Bought This Also Bought