Sentinel XL® OV/AG Cartridge Filters, ILC Dover
# de catalogue 76533-624
Fournisseur: ILC DOVER LP
Spécifications
- Description:OV/AG cartridge filters
- Cat. no.:76533-624
Spécifications
A propos de cet article
The OV/AG cartridge filters protect against particulates, organic vapors, chlorine, hydrogen chloride, sulfur dioxide, chlorine dioxide, and hydrogen fluoride.
- Multiple packaging options are available
Customers Who Bought This Also Bought