Sentinel XL® HEPA (HP) P100 Cartridge Filters, ILC Dover
# de catalogue 76533-616
Fournisseur: ILC DOVER LP
Spécifications
- Description:HEPA (HP P100 cartridge filters
- Cat. no.:76533-616
Spécifications
A propos de cet article
High efficiency particulate air cartridge filters can remove at least 99.97% of airborne particles with a size of 0.3 microns.
- Multiple packaging options are available
Use HEPA cartridge filters to eliminate airborne dust, pollen, mold, and bacteria.
Customers Who Bought This Also Bought