Sentinel XL® HP White Half Hood, ILC Dover
# de catalogue 76533-614
Fournisseur: ILC DOVER LP
Spécifications
- Description:Sentinel XL® HP whIte half hood
- Size:Universal
Spécifications
A propos de cet article
The Sentinel XL® half hood head cover offers an APF of 25.
- Hood provides a wide field of view
- Easily accommodates stethoscopes
Customers Who Bought This Also Bought