Sentinel XT® Clear Plus Full Hood, ILC Dover
# de catalogue 76533-606
Fournisseur: ILC DOVER LP
Spécifications
- Description:Sentinel XT® clear plus full hood
- Size:Universal
- Cat. no.:76533-606
Spécifications
A propos de cet article
The Sentinel XT® Clear Plus Full Hood is a superior solution when compared to other PAPR masks and facepieces.
- Ideal for use in the Pharmaceutical industry
- Designed for use with the Sentinel XT® Blower system
- 320 degree field of view
- No fit test required
- Save time and storage space with just one size
- APF of 1000
- Latex-free
With an APF of 1000, the Clear Plus Full Hood offers a 320 degree field of view, allowing the wearer to have facial hair and/or a head covering, and reducing feelings of claustrophobia. Wrap-around straps allow the outer bib to secure to protective clothing.
Customers Who Bought This Also Bought