Sentinel XL® HP Clear Full Hood Conversion Kit - CBRN to XL HP Clear Full Hood with 2 HEPA Filters, ILC Dover
# de catalogue 76533-556
Fournisseur: ILC DOVER LP
Spécifications
- Description:Sentinel XL® HP Clear full hood conversion kit
Spécifications
A propos de cet article
Convert the CBRN PAPR system into a HP PAPR System in a few easy steps.
- Go from an APF of 1000 to a NIOSH CAP2 protection rating in just minutes
- Save time and space, just one blower needed for CBRN and HP settings
- Easily change out hoods and filters
- Latex-free
The blower stays the same, only the hood and filters must be changed. Kit is supplied with 33" breathing tube, clear full hood, and 2 HEPA cartridge filters.
Customers Who Bought This Also Bought