Sentinel XL® Blower Kit, ILC Dover
# de catalogue 76533-550
Fournisseur: ILC DOVER LP
Spécifications
- Description:Sentinel XL® Blower kit
Spécifications
A propos de cet article
The Sentinel XL® Blower kit includes everything needed to get started.
Kit contains the Sentinel XL® Blower, NiMH rechargeable battery, charging cord, flow meter, Quick Loc belt, plug, and carrying tote. Cartridge filters, breathing tube, and hoods sold separately.
Customers Who Bought This Also Bought