Sentinel XL® HP Clear Half Hood System, ILC Dover
Catalog # 76533-544
Supplier: ILC DOVER LP
Specifications
- Description:Sentinel XL® HP Clear half hood system
- Size:Universal
- Cat. no.:76533-544
Specifications
About this item
Designed for use with Sentinel XL® blower, the Sentinel XL® HP clear half hood system is ideal for use in healthcare settings.
- No fit test required
- APF of 25
- Can be used easily with a stethoscope
- Available with or without duffel bag
- Available with HiMH rechargeable battery or alkaline battery pack
- Latex-free
Ideal for healthcare settings, the Sentinel XL® HP Clear Half Hood System includes the S-4001 clear half hood, 33" breathing tube, Sentinel XL Blower, flow meter, plug, NiMH rechargeable battery or alkaline battery pack, charging cord, Quick Loc belt, 2 HEPA filters, and carrying tote.
Customers Who Bought This Also Bought