Sentinel XL® Blower Assembly, ILC Dover
# de catalogue 76533-518
Fournisseur: ILC DOVER LP
Spécifications
- Description:Sentinel XL® blower assembly
Spécifications
A propos de cet article
The Sentinel XL® Blower has a rugged, sealed nylon protective shell, with floating polycarbonate internal blower assembly. The dual chamber design provides enhanced durability.
- Standard NIMH rechargeable battery pack can last up to 8 hours
- Alkaline battery pack can last up to 10 hours
- Blower and battery are lightweight and comfortable, weighing just 2.2 lbs.
The Sentinel XL® blower platform can be used in a variety of industries such as pharmaceutical, biopharmaceutical, infectious disease, along with chemical, biological, radiological, and nuclear applications. It can be used with Alkaline battery pack or a NiMH rechargeable battery.
Customers Who Bought This Also Bought