Spécifications
- Description:SCape® CBRN 30 escape respirator
- Size:Universal
- Cat. no.:76533-510
Spécifications
A propos de cet article
The SCape® CBRN30 from ILC Dover is the most advanced, FM and NIOSH-approved air-purifying escape respirator on the market. It provides mission critical protection from chemical, biological, radiological, and nuclear contaminants.
- Delivers reliable protection with minimal training
- Eases physiological burden on user
- Reduces claustrophobic feelings associated with other hoods
- Aids breathing for those with respiratory issues (e.g. Asthma)
A 'one size fits all' design eliminates the need for fit testing, as well as the need to store multiple sizes. The clear hood and large visor allows users to perform a wide range of tasks, including reading and driving. Additionally, this permits total facial recognition. The elimination of headbands and nose cups dramatically improves communication during an emergency, even with cell phones, radios, or other communication devices.
A NIOSH approved military grade canister provides protection against a range of military agents and NIOSH TRAs (filter data). A HEPA filter provides biological/radiological/particulate protection.