Masterflex® Standard Pump Head Tubing Loading Keys, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
Ensure proper tubing alignment and installation.
- Constructed of durable stainless steel
L/S® and I/P® Standard pump heads include tubing loading keys. A key is required to ensure proper tubing alignment and tension. Order a replacement key based on your pump head series, L/S® or I/P®.