Masterflex® Flow-Through Pump Tubing Weights, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
Keep your samples flowing and tubing from curling.
- Choose between PTFE or stainless steel construction
This flow-through tubing weight prevents tubing from curling or floating. Ideal for groundwater sampling, this weight fits Masterflex®; L/S® 15 or L/S® 24 Precision tubing.
Customers Who Bought This Also Bought