Masterflex® L/S® Variable-Speed Air-Powered Drive, 60 to 600 rpm
# de catalogue MFLX07569-00
Fournisseur: Avantor Fluid Handling
CE Compliant
Spécifications
- Control type:Analog Variable Speed
- Maximum Flow Rate:3400 ml/min
- Pump series:L/S
- Description:L/S variable speed air-powered drive, 60 to 600 rpm.
- Cat. no.:MFLX07569-00
- Max. RPM:600 rpm
- No. of Pump Heads Accepted:2
- Housing Material:Painted steel
- Drive height:224 mm (8.8125")
- Drive IP rating:IP 44
- Drive length:251 mm (9.875")
- Drive width:122 mm (4.8125")
- Min. RPM:60 rpm
- Motor hp:1/3
- Open-head sensor:No
- Reversible motor:No
- Speed control:±10%
- Type of pump heads accepted:Standard, Easy-Load, Easy-Load II, Quick Load, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm
- Motor:250 W
- Speed:Variable
- Warranty:2 years
Spécifications
A propos de cet article
Use where electrical power is unsafe or unavailable.
- Flow range 3.6 to 3400 ml/min—low rate depends on drive rpm, tubing head, and tubing size
- Easily vary speed by adjusting air pressure with regulator
- Rugged 1/3-hp air motor requires little maintenance and generates less heat than an electric motor
- IP44 rated—painted steel housing protects from gentle spray
Use this variable-speed air-powered drive whenever electrical power is unsafe or impractical. Careful grounding to protect from static electricity is vital for safe operation. Drive accepts nine pump head types: Standard, Easy-Load®, Easy-Load® II, Easy-Load® 3, Multichannel, High-Performance, Cartridge, PTFE-Tubing, and PTFE-Diaphragm.
Delivery: Includes air/speed regulator with 5-μm air filter, automatic lubricator, 0 to 30 psi pressure gauge, and muffler.
- Control type:Analog Variable Speed
- Maximum Flow Rate:3400 ml/min
- Pump series:L/S
- Description:L/S variable speed air-powered drive, 60 to 600 rpm.
- Cat. no.:MFLX07569-00
- Max. RPM:600 rpm
- No. of Pump Heads Accepted:2
- Housing Material:Painted steel
- Drive height:224 mm (8.8125")
- Drive IP rating:IP 44
- Drive length:251 mm (9.875")
- Drive width:122 mm (4.8125")
- Min. RPM:60 rpm
- Motor hp:1/3
- Open-head sensor:No
- Reversible motor:No
- Speed control:±10%
- Type of pump heads accepted:Standard, Easy-Load, Easy-Load II, Quick Load, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm
- Motor:250 W
- Speed:Variable
- Warranty:2 years
Customers Who Bought This Also Bought