FMI Series R Valveless Piston Pump Heads
Fournisseur: FLUID METERING INC
Certificats
A propos de cet article
Compatible with Masterflex® L/S® or series Q drive.
- No valves to clog or wear out
- Use for low-flow applications
- Kynar® pump body withstands harsh chemicals
- Series R pump heads come equipped with rotating pistons to prevent clogging—ideal for pumping visco
Compression fittings accept 1/4" OD tubing. For a complete assembly, order series R pump head and a Masterflex L/S® or series Q drive. Adapter kits let you attach pump heads to drives.
Flow characteristics may vary with different fluids.
Spécifications
- Model:
- 07117-99
- 07115-99
- 07117-67
- 07117-66
- Accuracy:
- ±1%
- ±1%
- Inlet connection:
- Series R Head to Series Q Drives Adapter
- Series R Head to L/S Drive Adapter
- 1/4" OD compression
- 1/4" OD compression
- Warranty:
- 1 year