Masterflex® Peristaltic Injector Pumps, Avantor®
Fournisseur: VWR International
Certificats
A propos de cet article
A cost-effective pump for pressure injection and chemical feed applications.
- Flow rates from 6 to 250 ml/min
- Valox® housing and tamper-resistant control cover
- High-pressure pumps are self priming
- Digital keypad with LED confirms pump settings and operation
- Built-in indicator alerts you when it is time to change tubing
- Variable-sp
With no internal check valves, these peristaltic pumps are not prone to clogging or fouling—ideal in applications with tough-to-handle solutions. Pump tubing assemblies last up to 1000 hours and take only minutes to change.
Delivery: Includes two Norprene® tubing assemblies, five feet of 3/8” OD vinyl suction tubing, five feet of 3/8” OD polyethylene discharge tubing, intake strainer with weight, and injector fitting with check valve.
Spécifications
- Model:
- 74201-00
- 74201-40
- 74201-20
- 74201-30
- 74201-10
- Duty cycle continuous:
- Yes
- Yes
- Yes
- Yes
- Yes
- ETL regulatory approval:
- Yes
- Yes
- Yes
- Yes
- Yes
- Inlet connection:
- 3/8" OD tubing
- 3/8" OD tubing
- 3/8" OD tubing
- 3/8" OD tubing
- 3/8" OD tubing
- Max. viscosity:
- 5000 cP
- 5000 cP
- 5000 cP
- 5000 cP
- 5000 cP
- Outlet connection:
- 3/8" OD tubing
- 3/8" OD tubing
- 3/8" OD tubing
- 3/8" OD tubing
- 3/8" OD tubing
- Wetted materials:
- Norprene, PVDF
- Norprene, PVDF
- Norprene, PVDF
- Norprene, PVDF
- Norprene, PVDF
- Max. pressure:
- 100 psi
- 50 psi
- 100 psi
- 50 psi
- 75 psi