Masterflex® Complete Drum and Container Pumps, Avantor®
À propos de cet article
System includes pump tube and motor—tube material options ensure chemical compatibility.
- Flow rate 35 GMP for all models
- Equipped with variable-speed control motors (except air and explosion-proof models) for added safety and control
- Maintenance is reduced with no mechanical seals
- Run dry capability
Drum and container pumps are engineered to safely transfer a variety of liquids from drums, tanks, and small storage vessels. Materials such as PP, PVDF, CPVC, and 316 SS provide excellent chemical resistance against corrosive liquids. Order optional dispensing nozzle from "Accessories" for added control.
Delivery information: Includes electric motor with 16-ft power cord and pump tube.
10 Options Available Below
Product Details & Documents
System includes pump tube and motor—tube material options ensure chemical compatibility.
- Flow rate 35 GMP for all models
- Equipped with variable-speed control motors (except air and explosion-proof models) for added safety and control
- Maintenance is reduced with no mechanical seals
- Run dry capability
Drum and container pumps are engineered to safely transfer a variety of liquids from drums, tanks, and small storage vessels. Materials such as PP, PVDF, CPVC, and 316 SS provide excellent chemical resistance against corrosive liquids. Order optional dispensing nozzle from "Accessories" for added control.
Delivery information: Includes electric motor with 16-ft power cord and pump tube.
Specifications for Product Family
Spécifications
- Catalog No:
- MFLX70613-00
- MFLX70613-05
- MFLX70613-10
- MFLX70613-15
- MFLX70613-20
- MFLX70613-25
- MFLX70613-30
- MFLX70613-35
- MFLX70613-40
- MFLX70613-45
- MFLX70613-50
- MFLX70613-55
- Model:
- 70613-00
- 70613-05
- 70613-10
- 70613-15
- 70613-20
- 70613-25
- 70613-30
- 70613-35
- 70613-40
- 70613-45
- 70613-50
- 70613-55
- CE regulatory approval:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. temperature:
- 55 °C (130 °F)
- 55 °C (130 °F)
- 55 °C (130 °F)
- 55 °C (130 °F)
- 80 °C (175 °F)
- 80 °C (175 °F)
- 80 °C (175 °F)
- 80 °C (175 °F)
- 90 °C (190 °F)
- 90 °C (190 °F)
- 90 °C (190 °F)
- 90 °C (190 °F)
- Max. viscosity:
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- Motor type:
- ODP
- ODP
- ODP
- ODP
- TEFC
- TEFC
- TEFC
- TEFC
- TEFC
- TEFC
- TEFC
- TEFC
- Motor hp:
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- Outlet connection:
- 1" hose barb
- 1" hose barb
- 1" hose barb
- 1" hose barb
- 1" hose barb
- 1" hose barb
- 1" hose barb
- 1" hose barb
- 1" hose barb
- 1" hose barb
- 1" hose barb
- 1" hose barb
- Tube length, inlet:
- 39"
- 39"
- 47"
- 47"
- 39"
- 39"
- 47"
- 47"
- 39"
- 39"
- 47"
- 47"
- UL US regulatory approval:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Wetted materials:
- Polypropylene, carbon, Hastelloy
- Polypropylene, carbon, Hastelloy
- Polypropylene, carbon, Hastelloy
- Polypropylene, carbon, Hastelloy
- PVDF, carbon, Hastelloy
- PVDF, carbon, Hastelloy
- PVDF, carbon, Hastelloy
- PVDF, carbon, Hastelloy
- CPVC, carbon, Hastelloy
- CPVC, carbon, Hastelloy
- CPVC, carbon, Hastelloy
- CPVC, carbon, Hastelloy
- Warranty:
- 3 years
- 3 years
- 3 years
- 3 years
- 3 years
- 3 years
- 3 years
- 3 years
- 3 years
- 3 years
- 3 years
- 3 years
Spécifications
Product Family Options
Informations sur le produit
- DésignationDébit maximalPuissanceMax. Motor RPMAvailabilityPrice
- Drum Pump, 35 gal./min, 39", PP, ODP, 110 VAC, 60 Hz132.49 L/min (35 gal./min)110 VAC; 60 Hz10000 rpmLog in pour la disponibilitéEach$2,360.00Spécifications:Intrinsicaly safeUL Listed
More
Specifications
Cat. No.MFLX70613-00Model70613-00CE regulatory approvalYesMax. temperature55 °C (130 °F)Max. viscosity1500 cPMotor typeODPMotor hp1Outlet connection1" hose barbTube length, inlet39"UL US regulatory approvalYesWetted materialsPolypropylene, carbon, HastelloyWarranty3 yearsIntrinsicaly safeUL Listed - Drum Pump, 35 gal./min, 39", PP, ODP, 230 VAC, 50 Hz132.49 L/min (35 gal./min)230 VAC; 50 Hz10000 rpmLog in pour la disponibilitéEach$2,590.00Spécifications:CE CompliantIntrinsicaly safeUL Listed
More
Specifications
Cat. No.MFLX70613-05Model70613-05CE regulatory approvalYesMax. temperature55 °C (130 °F)Max. viscosity1500 cPMotor typeODPMotor hp1Outlet connection1" hose barbTube length, inlet39"UL US regulatory approvalYesWetted materialsPolypropylene, carbon, HastelloyWarranty3 yearsCE CompliantIntrinsicaly safeUL Listed - Drum Pump, 35 gal./min, 47", PP, ODP, 110 VAC, 60 Hz132.49 L/min (35 gal./min)110 VAC; 60 Hz10000 rpmLog in pour la disponibilitéEach$2,540.00Spécifications:Intrinsicaly safeUL Listed
More
Specifications
Cat. No.MFLX70613-10Model70613-10CE regulatory approvalYesMax. temperature55 °C (130 °F)Max. viscosity1500 cPMotor typeODPMotor hp1Outlet connection1" hose barbTube length, inlet47"UL US regulatory approvalYesWetted materialsPolypropylene, carbon, HastelloyWarranty3 yearsIntrinsicaly safeUL Listed - Drum Pump, 35 gal./min, 47", PP, ODP, 230 VAC, 50 Hz132.49 L/min (35 gal./min)230 VAC; 50 Hz10000 rpmDiscontinued-Spécifications:CE CompliantIntrinsicaly safeUL Listed
More
Specifications
Cat. No.MFLX70613-15Model70613-15CE regulatory approvalYesMax. temperature55 °C (130 °F)Max. viscosity1500 cPMotor typeODPMotor hpOutlet connection1" hose barbTube length, inlet47"UL US regulatory approvalYesWetted materialsPolypropylene, carbon, HastelloyWarranty3 yearsCE CompliantIntrinsicaly safeUL Listed - Drum Pump, 35 gal./min, 39", PVDF, TEFC, 110 VAC, 60 Hz132.49 L/min (35 gal./min)110 VAC; 60 Hz10000 rpmLog in pour la disponibilitéEach$3,640.00Spécifications:Intrinsicaly safeUL Listed
More
Specifications
Cat. No.MFLX70613-20Model70613-20CE regulatory approvalYesMax. temperature80 °C (175 °F)Max. viscosity1500 cPMotor typeTEFCMotor hp1Outlet connection1" hose barbTube length, inlet39"UL US regulatory approvalYesWetted materialsPVDF, carbon, HastelloyWarranty3 yearsIntrinsicaly safeUL Listed - Drum Pump, 35 gal./min, 39", PVDF, TEFC, 230 VAC, 50 Hz132.49 L/min (35 gal./min)230 VAC; 50 Hz10000 rpmLog in pour la disponibilitéEach$3,730.00Spécifications:CE CompliantIntrinsicaly safeUL Listed
More
Specifications
Cat. No.MFLX70613-25Model70613-25CE regulatory approvalYesMax. temperature80 °C (175 °F)Max. viscosity1500 cPMotor typeTEFCMotor hp1Outlet connection1" hose barbTube length, inlet39"UL US regulatory approvalYesWetted materialsPVDF, carbon, HastelloyWarranty3 yearsCE CompliantIntrinsicaly safeUL Listed - Drum Pump, 35 gal./min, 47", PVDF, TEFC, 110 VAC, 60 Hz132.49 L/min (35 gal./min)110 VAC; 60 Hz10000 rpmDiscontinued-Spécifications:Intrinsicaly safeUL Listed
More
Specifications
Cat. No.MFLX70613-30Model70613-30CE regulatory approvalYesMax. temperature80 °C (175 °F)Max. viscosity1500 cPMotor typeTEFCMotor hp1Outlet connection1" hose barbTube length, inlet47"UL US regulatory approvalYesWetted materialsPVDF, carbon, HastelloyWarranty3 yearsIntrinsicaly safeUL Listed - Drum Pump, 35 gal./min, 47", PVDF, TEFC, 230 VAC, 50 Hz132.49 L/min (35 gal./min)230 VAC; 50 Hz10000 rpmLog in pour la disponibilitéEach$3,800.00Spécifications:CE CompliantIntrinsicaly safeUL Listed
More
Specifications
Cat. No.MFLX70613-35Model70613-35CE regulatory approvalYesMax. temperature80 °C (175 °F)Max. viscosity1500 cPMotor typeTEFCMotor hp1Outlet connection1" hose barbTube length, inlet47"UL US regulatory approvalYesWetted materialsPVDF, carbon, HastelloyWarranty3 yearsCE CompliantIntrinsicaly safeUL Listed - Drum Pump, 35 gal./min, 39", CPVC, TEFC, 110 VAC, 60 Hz132.49 L/min (35 gal./min)110 VAC; 60 Hz10000 rpmLog in pour la disponibilitéEach$3,140.00Spécifications:Intrinsicaly safeUL Listed
More
Specifications
Cat. No.MFLX70613-40Model70613-40CE regulatory approvalYesMax. temperature90 °C (190 °F)Max. viscosity1500 cPMotor typeTEFCMotor hp1Outlet connection1" hose barbTube length, inlet39"UL US regulatory approvalYesWetted materialsCPVC, carbon, HastelloyWarranty3 yearsIntrinsicaly safeUL Listed - Drum Pump, 35 gal./min, 39", CPVC, TEFC, 230 VAC, 50 Hz132.49 L/min (35 gal./min)230 VAC; 50 Hz10000 rpmLog in pour la disponibilitéEach$3,220.00Spécifications:CE CompliantIntrinsicaly safeUL Listed
More
Specifications
Cat. No.MFLX70613-45Model70613-45CE regulatory approvalYesMax. temperature90 °C (190 °F)Max. viscosity1500 cPMotor typeTEFCMotor hp1Outlet connection1" hose barbTube length, inlet39"UL US regulatory approvalYesWetted materialsCPVC, carbon, HastelloyWarranty3 yearsCE CompliantIntrinsicaly safeUL Listed - Drum Pump, 35 gal./min, 47", CPVC, TEFC, 110 VAC, 60 Hz132.49 L/min (35 gal./min)110 VAC; 60 Hz10000 rpmLog in pour la disponibilitéEach$2,940.00Spécifications:Intrinsicaly safeUL Listed
More
Specifications
Cat. No.MFLX70613-50Model70613-50CE regulatory approvalYesMax. temperature90 °C (190 °F)Max. viscosity1500 cPMotor typeTEFCMotor hp1Outlet connection1" hose barbTube length, inlet47"UL US regulatory approvalYesWetted materialsCPVC, carbon, HastelloyWarranty3 yearsIntrinsicaly safeUL Listed - Drum Pump, 35 gal./min, 47", CPVC, TEFC, 230 VAC, 50 Hz132.49 L/min (35 gal./min)230 VAC; 50 Hz10000 rpmLog in pour la disponibilitéEach$3,240.00Spécifications:CE CompliantIntrinsicaly safeUL Listed
More
Specifications
Cat. No.MFLX70613-55Model70613-55CE regulatory approvalYesMax. temperature90 °C (190 °F)Max. viscosity1500 cPMotor typeTEFCMotor hp1Outlet connection1" hose barbTube length, inlet47"UL US regulatory approvalYesWetted materialsCPVC, carbon, HastelloyWarranty3 yearsCE CompliantIntrinsicaly safeUL Listed
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement Barrel Adapter for Drum Pumps, PPReplacement Barrel Adapter for Drum Pumps, PPMFLX70604-92Log in pour la disponibilitéEach - $97.90
Informations sur le produit
- Catalog #Motor TypePowerAvailabilityPrice
- Replacement motor, ODP, 110 VAC, 60 HzReplacement motor, ODP, 110 VAC, 60 HzMFLX70604-03Open drip proof (ODP)110 VAC; 60 HzLog in pour la disponibilitéEach - $1,450.00
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement Wall Mounting Bracket for Drum PumpsReplacement Wall Mounting Bracket for Drum PumpsMFLX70604-90Log in pour la disponibilitéEach - $99.60
Informations sur le produit
- Catalog #AvailabilityPrice
- Drum Pump Tube Set, PP, 39" L
Informations sur le produit
- Catalog #MaterialAvailabilityPrice
- Strainer for Drum Pumps, PP
Informations sur le produit
- Catalog #AvailabilityPrice
- Drum Pump Tube Set, PP, 47"L
Informations sur le produit
- Catalog #AvailabilityPrice
- Drum Pump Tube Set, PVDF, 39"
Informations sur le produit
- Catalog #Motor TypePowerAvailabilityPrice
- Replacement motor, TEFC, 110 VAC, 60 HzReplacement motor, TEFC, 110 VAC, 60 HzMFLX70604-13Totally enclosed, fan cooled (TEFC)110 VAC; 60 HzLog in pour la disponibilitéEach - $1,890.00
Informations sur le produit
- Catalog #AvailabilityPrice
- Dispensing Nozzle, PVDF, with 6-ft of PVC hoseDispensing Nozzle, PVDF, with 6-ft of PVC hoseMFLX70604-98Log in pour la disponibilitéEach - $1,470.00
Informations sur le produit
- Catalog #AvailabilityPrice
- Drum Pump Tube Set, PVDF, 47"
Informations sur le produit
- Catalog #AvailabilityPrice
- Drum Pump Tube Set, CPVC, 39"L
Informations sur le produit
- Catalog #Motor TypePowerAvailabilityPrice
- Replacement motor, TEFC, 230 VAC, 50 HzReplacement motor, TEFC, 230 VAC, 50 HzMFLX70604-16Totally enclosed fan cooled (TEFC)230 VAC; 50 HzLog in pour la disponibilitéEach - $2,020.00
Informations sur le produit
- Catalog #AvailabilityPrice
- Drum Pump Tube Set, CPVC, 47"L