Masterflex® Transfer Tubing, Fluorinated Etyhlene Propylene (FEP), Avantor®
Fournisseur: VWR International
Certificats
A propos de cet article
Does not deteriorate with age.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Excellent for the transfer of extreme temperature or highly corrosive materials. Also good for high-purity work.
Characteristics: Chemically inert, nontoxic, heat resistant, with a low coefficient of friction. Doesn’t deteriorate with age. Tough, but bends even at -454 °F (-270 °C). Translucent.
Certifications: Resin is FDA-compliant (21 CFR 177.1550). Tubing is manufactured in ISO 9001-2008 approved facility.
Temperature range: -454 to 400 °F (-270 to 205 °C)
Sterilize: Autoclave, ethylene oxide, or use dry heat.
Spécifications
- Catalog No:
- MFLX06406-60
- MFLX06406-62
- MFLX06450-01
- MFLX06450-14
- MFLX06406-74
- MFLX06450-08
- MFLX06406-76
- MFLX06450-09
- MFLX06450-15
- MFLX06406-12
- MFLX06450-17
- MFLX06406-63
- MFLX06450-04
- MFLX06406-64
- MFLX06450-05
- MFLX06406-66
- MFLX06450-06
- MFLX06406-72
- MFLX06450-07
- MFLX06406-18
- MFLX06450-19
- MFLX06450-00
- MFLX06450-02
- Model:
- 06406-60
- 06406-62
- 06450-01
- 06450-14
- 06406-74
- 06450-08
- 06406-76
- 06450-09
- 06450-15
- 06406-12
- 06450-17
- 06406-63
- 06450-04
- 06406-64
- 06450-05
- 06406-66
- 06450-06
- 06406-72
- 06450-07
- 06406-18
- 06450-19
- 06450-00
- 06450-02
- GMP compliant:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Length per pack:
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- Max. pressure:
- 34.5 bar (500 psi)
- 31.6 bar (444 psi)
- 4.5 bar (65 psi)
- 12.8 bar (185 psi)
- 9.7 bar (140 psi)
- 16.8 bar (243 psi)
- 7 bar (102 psi)
- 14.5 bar (210 psi)
- 11.4 bar (165 psi)
- 7.6 bar (110 psi)
- 9.3 bar (135 psi)
- 24.3 bar (353 psi)
- 22.7 bar (329 psi)
- 19.9 bar (289 psi)
- 31 bar (450 psi)
- 14.4 bar (209 psi)
- 24.3 bar (353 psi)
- 10.3 bar (150 psi)
- 19.9 bar (288 psi)
- 6.1 bar (88 psi)
- 7.9 bar (115 psi)
- 4.9 bar (71 psi)
- 11.9 bar (172 psi)
- Max. temperature:
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- 204 °C (400 °F)
- Regulatory approvals:
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- Sterilisation method autoclave:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method EtO:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes