Masterflex® B/T® PerfectPosition® Pump Tubing, Peroxide-Cured Silicone, Avantor®
Fournisseur: VWR International
Certificats
A propos de cet article
Ensure optimum performance from your Masterflex® pump.
- Excellent biocompatibility
- No additives, plasticizers or DOP; odorless and nontoxic, fungus-resistant
- No taste imparted to transported fluids
- Extremely good at low temperatures
- Weather, ozone, corona, and radiation resistant
- Minimal tendency to take a set
- PerfectPosition™ tubing retention marks indicate the exact placement of tubing in the pump head to provide the best performance and life of the tubing
- Custom extruded to fit 77110- and 77111-series Masterflex® B/T® pumps and pump heads
- Engineered for long life in peristaltic pump applications
Characteristics: Superior heat resistance, low compression set, odorless, and nontoxic. Translucent.
Certifications: Elastomer is FDA-compliant (21 CFR 177.2600). Tubing is
manufactured according to GMP.
Temperature range: –60 to 460 °F (–51 to 238 °C)
Sterilize: Autoclave or gamma irradiation.
Spécifications
- Catalog No:
- MFLX96400-91
- MFLX96400-90
- Model:
- 96400-91
- 96400-90
- Length per pack:
- 3 m (10 ft.)
- 3 m (10 ft.)
- Max. pressure:
- 2.06 bar (30 psi)
- 2.4 bar (35 psi)
- Max. temperature:
- 230 °C (446 °F)
- 230 °C (446 °F)
- Min. temperature (dynamic):
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- Max. temperature (dynamic):
- 100 °C (212 °F)
- 100 °C (212 °F)
- Pump series:
- B/T
- B/T
- Regulatory approvals:
- 3A;ADCF;European Pharmacopoeia;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
- 3A;ADCF;European Pharmacopoeia;FDA;Non-DEHP;Non-Phthalate;RoHS;USP