Masterflex® Storage Racks for Fitting Kits, Avantor®
Fournisseur: VWR International
Certificats
A propos de cet article
Organize fittings for easy access.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Organize your workspace with these Masterflex® fitting kit storage racks. Storage racks hold up to four fitting kits. Simply slide the complete fitting kit into the rack for secure storage. Storage racks are for kits only, not assortment packs. Fitting kit with fittings are sold separately.
Customers Who Bought This Also Bought