Masterflex® Liquid Flow Switches for Threaded Plastic Piping, Avantor®
À propos de cet article
Connects quickly to threaded plastic piping.
- Chemically resistant — wetted materials stainless steel and polypropylene
- Effortlessly install with 1/2" NPT(M) connections
- NSF and FDA approved
These normally open switches have 3/4" NPT(F) ports for a quick connection to threaded plastic piping. Polypropylene and stainless steel wetted parts offer excellent chemical resistance. The materials are also NSF approved and FDA compliant; approved for potable water treatment applications such as chlorinators, purifiers, and heaters. Switches include 24" lead wires.
5 Options Available Below
Product Details & Documents
Connects quickly to threaded plastic piping.
- Chemically resistant — wetted materials stainless steel and polypropylene
- Effortlessly install with 1/2" NPT(M) connections
- NSF and FDA approved
These normally open switches have 3/4" NPT(F) ports for a quick connection to threaded plastic piping. Polypropylene and stainless steel wetted parts offer excellent chemical resistance. The materials are also NSF approved and FDA compliant; approved for potable water treatment applications such as chlorinators, purifiers, and heaters. Switches include 24" lead wires.
Specifications for Product Family
Spécifications
- Model:
- 32774-60
- 32774-62
- 32774-64
- 32774-66
- 32774-68
- Max. pressure:
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- Max. operating temperature:
- 100 °C (212 °F)
- 100 °C (212 °F)
- 100 °C (212 °F)
- 100 °C (212 °F)
- 100 °C (212 °F)
- Wetted materials:
- polypropylene, Viton®, 316 SS, and PH 15-7 Mo stainless steel
- polypropylene, Viton®, 316 SS, and PH 15-7 Mo stainless steel
- polypropylene, Viton®, 316 SS, and PH 15-7 Mo stainless steel
- polypropylene, Viton®, 316 SS, and PH 15-7 Mo stainless steel
- polypropylene, Viton®, 316 SS, and PH 15-7 Mo stainless steel
- Process connection:
- 0.75 NPT(F)
- 0.75 NPT(F)
- 0.75 NPT(F)
- 0.75 NPT(F)
- 0.75 NPT(F)
- Height:
- 40749"
- 40749"
- 40749"
- 40749"
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Spécifications
Product Family Options
Informations sur le produit
- Débit maximalDésignationAvailabilityPrice
- 0.25 gal./minLiquid flow switch for threaded plastic piping; 0,25 gal./minLog in pour la disponibilitéEach$294.00Spécifications:NSF international certified
More
Specifications
Cat. No.MFLX32774-60Model32774-60Max. pressure100 psiMax. operating temperature100 °C (212 °F)Wetted materialspolypropylene, Viton®, 316 SS, and PH 15-7 Mo stainless steelProcess connection0.75 NPT(F)Height40749"Warranty1 yearNSF international certified - 0.5 gal./minLiquid flow switch for threaded plastic piping; 0,5 gal./minLog in pour la disponibilitéEach$314.00Spécifications:NSF international certified
More
Specifications
Cat. No.MFLX32774-62Model32774-62Max. pressure100 psiMax. operating temperature100 °C (212 °F)Wetted materialspolypropylene, Viton®, 316 SS, and PH 15-7 Mo stainless steelProcess connection0.75 NPT(F)HeightWarranty1 yearNSF international certified - 1 gal./minLiquid flow switch for threaded plastic piping; 1,0 gal./minLog in pour la disponibilitéEach$294.00Spécifications:NSF international certified
More
Specifications
Cat. No.MFLX32774-64Model32774-64Max. pressure100 psiMax. operating temperature100 °C (212 °F)Wetted materialspolypropylene, Viton®, 316 SS, and PH 15-7 Mo stainless steelProcess connection0.75 NPT(F)Height40749"Warranty1 yearNSF international certified - 2 gal./minLiquid flow switch for threaded plastic piping; 2,0 gal./minLog in pour la disponibilitéEach$294.00Spécifications:NSF international certified
More
Specifications
Cat. No.MFLX32774-66Model32774-66Max. pressure100 psiMax. operating temperature100 °C (212 °F)Wetted materialspolypropylene, Viton®, 316 SS, and PH 15-7 Mo stainless steelProcess connection0.75 NPT(F)Height40749"Warranty1 yearNSF international certified - 2.5 gal./minLiquid flow switch for threaded plastic piping; 2,5 gal./minLog in pour la disponibilitéEach$294.00Spécifications:NSF international certified
More
Specifications
Cat. No.MFLX32774-68Model32774-68Max. pressure100 psiMax. operating temperature100 °C (212 °F)Wetted materialspolypropylene, Viton®, 316 SS, and PH 15-7 Mo stainless steelProcess connection0.75 NPT(F)Height40749"Warranty1 yearNSF international certified