Masterflex® High-Efficiency Absolute-Rated Filter Bags, Polypropylene, Avantor®
Supplier: PENTAIR RESIDENTIAL FILTRATION LLC
Certificates
About this item
Wide range of options for temperature, flow rate, and filtration quality.
- Constructed of polypropylene (PP)
- FDA-compliant
Designed for 98% removal efficiency at the rated pore size, these bags offer three to four times the dirt-holding capacity of standard bags. In addition, they are FDA-compliant with respect to all regulations governing food-grade materials. These polypropylene absolute-rated filter bags are rated for a temperature of 200 °F (93 °C) maximum.
Specifications
- Filter diameter:
- 7"
- 7"
- 7"
- 7"
- Filtration area:
- 0.409 m² (4.4 ft.², 633.6 in²)
- 0.409 m² (4.4 ft.², 633.6 in²)
- 0.409 m² (4.4 ft.², 633.6 in²)
- 0.409 m² (4.4 ft.², 633.6 in²)
- Max. temperature:
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- Pore size:
- 1 µm
- 100 µm
- 25 µm
- 50 µm
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year