Masterflex® L/S® Digital Miniflex® Pump Systems, Avantor®
Fournisseur: Avantor Fluid Handling
Certificats
A propos de cet article
Make low-flow pumping easy!
- Flow range 0.004 to 315 ml/min—flow rate depends on drive rpm, pump head, and tubing size
- Digital interface with graphical LCD shows pump performance and operating mode
- Remote control available using analog communication—voltage, current and contact closure
- Integrated “open-head” sensor halts operation when pump head is opened
- Pump head accepts continuous lengths of tubing for clean flow paths
- Automatic tubing retention makes tube loading quick and easy
- More than 20% smaller than competitive units—compact size and light weight make it easy to move and share between labs
This compact, stackable pump features a durable ABS housing that is easy to wipe down and keep clean. The Miniflex pump head features a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing. Self-lubricating thermoplastic polyester rollers are low friction and long wearing for smooth operation and long life.
Note:Dual-channel model must have same size tubing loaded in both channels to ensure tubing retention.
Delivery: Includes l/S Miniflex pump head and universal voltage (90 to 260 VAC) benchtop power supply with IEC320/CEE22 socket. Pump is shipped with cord/plug assembly specific to country of destination.
Spécifications
- Catalog No:
- MFLX07525-40
- MFLX07525-20
- No. of channels:
- 2
- 1
- Control type:
- Variable-Speed Digital
- Digital
- Min. flow rate:
- 0.008 ml/min
- 0.004 ml/min
- Accuracy:
- ±0.1%
- ±0.1%
- Cloud connected:
- No
- No
- Pump series:
- L/S
- L/S
- Frequency:
- 50/60 Hz
- 50/60 Hz