Masterflex® L/S® Variable-Speed Economy Modular Drive Systems, Avantor®
Fournisseur: Avantor Fluid Handling
Certificats
A propos de cet article
Versatile variable-speed pump system with a wide flow range and simple controls.
- Separate motor and controller are connected via a 6-ft (1.9 m) cable — place components where convenient or where space allows
- Single-turn speed control and on/off switch with green power indicator — maintain speed setting while turning pump on/off
- The 1/10-hp reversible motor has soft start and back EMF for ±2% speed control
- Painted steel housing is IP33 rated for protection from vertically falling water
Choose a convenient, complete system all with one catalog number. A perfect upgrade from a fixed-speed drive, these variable-speed drives offer a wider rpm range and ±2% speed regulation.
Spécifications
- No. of channels:
- 1
- 1
- 1
- 1
- Control type:
- Variable-Speed Analog
- Variable-Speed Analog
- Variable-Speed Analog
- Variable-Speed Analog
- Min. flow rate:
- 56 ml/min
- 4.8 ml/min
- 56 ml/min
- 4.8 ml/min
- Accuracy:
- ±2%
- ±2%
- ±2%
- ±2%
- Cloud connected:
- No
- No
- No
- No
- Pump series:
- L/S
- L/S
- L/S
- L/S
- Frequency:
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz