Masterflex® L/S® Digital Modular Drives Replacement Parts, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
Make the most of your Masterflex drives replacement parts and other accessories.
- Extend the life of your Masterflex pump with these replacement parts
Choose replacement motors based on your Masterflex Modular System.
Specifications
- Catalog No:
- MFLX77301-52
- MFLX77301-54
- MFLX77301-44
- MFLX77301-42
- MFLX77301-64
- MFLX77301-62
- MFLX77301-72
- MFLX77301-74
- Model:
- 77301-52
- 77301-54
- 77301-44
- 77301-42
- 77301-72
- 77301-74
- Controller height:
- 119 mm (4.75")
- 140 mm (5.5")
- 140 mm (5.5")
- 119 mm (4.75")
- Controller length:
- 285 mm (10.2031")
- 263 mm (10.4062")
- 263 mm (10.4062")
- 285 mm (10.2031")
- Controller width:
- 279 mm (11")
- 211 mm (8.4062")
- 211 mm (8.4062")
- 279 mm (11")
- Motor:
- 75 W
- 75 W
- 75 W
- 75 W
- Motor hp:
- 1/10
- 1/10
- 1/10
- 1/10
- Pump series:
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- Speed control:
- ±0.1%
- ±0.1%
- ±0.1%
- ±0.1%
- Controller IP rating:
- IP 66
- IP 33
- IP 33
- IP 66
- Motor height:
- 152 mm (6")
- 117 mm (4.6093")
- 117 mm (4.6093")
- 152 mm (6")
- Motor IP rating:
- IP 66
- IP 33
- IP 33
- IP 66
- Motor length:
- 395 mm (15.6093")
- 267 mm (10.5")
- 267 mm (10.5")
- 395 mm (15.6093")
- Motor width:
- 178 mm (7")
- 97 mm (3.7968")
- 97 mm (3.7968")
- 178 mm (7")
- Warranty:
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years