Masterflex® L/S® Multichannel Pump Head Cartridges, Avantor®
Fournisseur: Avantor Fluid Handling
Certificats
A propos de cet article
Use with L/S® multichannel cartridge pump heads for synchronous, multichannel pumping.
- Only Silicone and C-Flex® tubing formulations are recommended for size L/S® 17 tubing when used in L/S® cartridge pump heads.
Cartridges feature accurate flow rates and adjustable tubing retainers to hold tubing in place — for large cartridges, set tubing retainers to your tube size and you are ready to pump. On small cartridges, optimum retention is preset — simply load tubing and start pumping.
Cartridges are constructed from polycarbonate and glass-filled nylon. Mix small and large cartridges on pump heads that accept both sizes; two small cartridges occupy the same space as one large cartridge.
Spécifications
- Model:
- 07519-70
- 07519-80
- 07519-75
- 07519-85
- No. of channels:
- 1
- 1
- 1
- 1
- Min. flow rate per channel:
- 0.004 ml/min
- 0.0015 ml/min
- 0.0006 ml/min
- 1E-05 ml/min
- Max. flow rate per channel:
- 1700 ml/min
- 130 ml/min
- 350 ml/min
- 85 ml/min
- Open-head sensor:
- No
- No
- No
- No
- Pump series:
- L/S
- L/S
- L/S
- L/S
- Occlusion:
- Adjustable Tangential
- Adjustable Tangential
- Adjustable Offset Tangential
- Adjustable Offset Tangential