Masterflex® Transfer Tubing, Premo Flex®, Avantor®
Supplier: VWR International
Certificates
About this item
Resists weather and ozone.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Outstanding resistance to heat and ozone
Masterflex® Premp Flex® transfer tubing is ideal for use with fuels such as petroleum oils, gasoline, kerosene, and lubricating oils, while maintaining tube integrity. The Buna-N tube, modified Buna-N nitrile tube cover, along with the high tensile textile core tube reinforcement makes this tubing suitable for high pressure applications. The tube meets RMA Class A high oil resistance and nonconductive at 1000 VDC certification. The tube cover meets Class B medium oil resistance and nonconductive at 1000 VDC certification.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.
Specifications
- Model:
- 95953-07
- 95953-01
- 95953-05
- 95953-03
- Bend radius:
- 203.2 mm (8")
- 50.8 mm (2")
- 127 mm (5")
- 101.6 mm (4")
- Clarity:
- Opaque
- Opaque
- Opaque
- Opaque
- Length per pack:
- 15.2 m (50 ft.)
- 30.5 m (100 ft.)
- 15.2 m (50 ft.)
- 30.5 m (100 ft.)
- Max. pressure:
- 17.2 bar (250 psi)
- 17.2 bar (250 psi)
- 17.2 bar (250 psi)
- 17.2 bar (250 psi)
- Max. temperature:
- 100 °C
- 100 °C
- 100 °C
- 100 °C
- Min. temperature:
- –28 °C (–20 °F)
- –28 °C (–20 °F)
- –28 °C (–20 °F)
- –28 °C (–20 °F)
- Sterilisation method autoclave:
- No
- No
- No
- No
- Sterilisation method EtO:
- No
- No
- No
- No
- Sterilisation method gamma:
- No
- No
- No
- No
- Working pressure:
- 17.2 bar (250 psi)
- 17.2 bar (250 psi)
- 17.2 bar (250 psi)
- 17.2 bar (250 psi)
- Weight per ft.:
- 0.24 kg (0.54 lbs.)
- 0.04 kg (0.09 lbs.)
- 0.16 kg (0.36 lbs.)
- 0.1 kg (0.22 lbs.)